1,2,3,4-Tetrahydrobenzo[b]azepin-5-one 97%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydro-benzo[b]azepin-5-one, 1,2,3,4-Tetrahydrobenzazepine-5-one, 2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

A193685-100MG A193685-10G A193685-1G A193685-250MG A193685-25G A193685-5G
76690-960EA 5.31 USD
76690-960 76690-962 76690-964 76690-966 76690-968 76690-970
1,2,3,4-Tetrahydrobenzo[b]azepin-5-one 97%
1,2,3,4-Tetrahydrobenzo[b]azepin-5-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MW: 161.204 g/mol
MDL Number: MFCD03426404
CAS Number: 1127-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR