5-Nitro-8-quinolinol 98%

Supplier: Ambeed
Danger

A193247-100MG A193247-1G A193247-25G A193247-5G
76690-952EA 5 USD
76690-952 76690-954 76690-956 76690-958
5-Nitro-8-quinolinol 98%
5-Nitro-8-quinolinol

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD00006791
CAS Number: 4008-48-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR