1,3-Benzoxazole-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A192804-100MG A192804-1G A192804-250MG A192804-5G A192804-25G
76690-944EA 9 USD
76690-944 76690-946 76690-948 76690-950 77877-437
1,3-Benzoxazole-6-carboxylic acid 97%
1,3-Benzoxazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₃
MDL Number: MFCD09954949
CAS Number: 154235-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR