Bis(4-bromophenyl)ether 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dibromodiphenyl Ether, 4,4'-Oxybis(bromobenzene), 4 4'-OXYBIS(BROMOBENZENE)

A190801-100G A190801-25G A190801-500G A190801-5G
76690-868EA 39 USD
76690-868 76690-870 76690-872 76690-874
Bis(4-bromophenyl)ether 98%
Bis(4-bromophenyl)ether

Precaución: for Research Use Only

Formula: C₁₂H₈Br₂O
MDL Number: MFCD00000095
CAS Number: 2050-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR