7-Bromo-4-fluoroindole 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-4-fluoro-1H-indole

A190341-100MG A190341-10G A190341-1G A190341-250MG A190341-5G
76690-856EA 51.99 USD
76690-856 76690-858 76690-860 76690-862 76690-864
7-Bromo-4-fluoroindole 98%
7-Bromo-4-fluoroindole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.04 g/mol
MDL Number: MFCD04037874
CAS Number: 292636-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR