3'-Iodoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Iodophenyl)ethanone, m-Iodoacetophenone, 1-(3-Iodophenyl)ethan-1-one

A190182-100G A190182-10G A190182-1G A190182-25G A190182-500G A190182-5G
76690-838EA 370.26 USD
76690-838 76690-840 76690-842 76690-844 76690-846 76690-848
3'-Iodoacetophenone 98%
3'-Iodoacetophenone

Precaución: for Research Use Only

Formula: C₈H₇IO
MW: 246.05 g/mol
MDL Number: MFCD00060283
CAS Number: 14452-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR