3,5-Xylylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dimethylphenyl)acetic acid, (3,5-Dimethylphenyl)acetic acid

A190141-5G A190141-1G
76690-836EA 162.32 USD
76690-836 77600-544
3,5-Xylylacetic acid 98%
3,5-Xylylacetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164.2 g/mol
MDL Number: MFCD00082776
CAS Number: 42288-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR