Delapril hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanyl-N-(2,3-dihydroinden-2-yl)glycine Hydrochloride, 2-((S)-N-(2,3-Dihydro-1H-inden-2-yl)-2-(((S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanamido)acetic acid hydrochloride, Delapril HCl

A189764-10MG A189764-25MG A189764-50MG A189764-5MG A189764-1MG
76690-828EA 8 USD
76690-828 76690-830 76690-832 76690-834 77600-540
Delapril hydrochloride 97%
Delapril hydrochloride

Precaución: for Research Use Only

Formula: C₂₆H₃₂N₂O₅
MW: 489.01 g/mol
MDL Number: MFCD00884619
CAS Number: 83435-67-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR