(S)-3-((tert-Butoxycarbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid 97%

Supplier: Ambeed
Warning

A188559-100MG A188559-250MG
76690-774EA 12.74 USD
76690-774 76690-776
(S)-3-((tert-Butoxycarbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid 97%
(S)-3-((tert-Butoxycarbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.19 g/mol
MDL Number: MFCD03427904
CAS Number: 499995-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR