7-Bromochroman-4-one 97%

Supplier: Ambeed
Warning

A187395-100G A187395-10G A187395-1G A187395-250MG A187395-25G A187395-5G
76690-736EA 1076.82 USD
76690-736 76690-738 76690-740 76690-742 76690-744 76690-746
7-Bromochroman-4-one 97%
7-Bromochroman-4-one

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.0600 g/mol
MDL Number: MFCD09744055
CAS Number: 18442-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR