Ethyl-2,5-dichlorobenzoate 98%

Supplier: Ambeed
Warning

A187089-100G A187089-25G A187089-5G
76690-722EA 64 USD
76690-722 76690-724 76690-726
Ethyl-2,5-dichlorobenzoate 98%
Ethyl-2,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MDL Number: MFCD00013636
CAS Number: 35112-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR