Anthraquinone-2,7-disulfonic acid disodium salt 80%

Supplier: Ambeed
Warning

Synonyms: Sodium 9,10-dioxo-9,10-dihydroanthracene-2,7-disulphonate, Disodium Anthraquinone-2,7-disulphonate

A186175-100G A186175-10G A186175-1G A186175-25G A186175-500G A186175-5G
76690-704EA 286 USD
76690-704 76690-706 76690-708 76690-710 76690-712 76690-714
Anthraquinone-2,7-disulfonic acid disodium salt 80%
Anthraquinone-2,7-disulfonic acid disodium salt

Precaución: for Research Use Only

Formula: C₁₄H₈O₈S₂
MDL Number: MFCD00053255
CAS Number: 853-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR