tert-Butyl (1,2,3,4-tetrahydroquinolin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A184552-250MG A184552-5G
76690-668EA 62 USD
76690-668 77877-350
tert-Butyl (1,2,3,4-tetrahydroquinolin-3-yl)carbamate 97%
tert-Butyl (1,2,3,4-tetrahydroquinolin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD02181058
CAS Number: 219862-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR