4-Fluoroisatoic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione

A184231-1G A184231-250MG A184231-5G
76690-662EA 40.32 USD
76690-662 76690-664 76690-666
4-Fluoroisatoic anhydride 97%
4-Fluoroisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄FNO₃
MW: 181.1200 g/mol
MDL Number: MFCD03839826
CAS Number: 321-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR