ω-Laurinlactam 99%

Supplier: Ambeed
Warning

Synonyms: Cyclododecanone Isooxime, Azacyclotridecan-2-one

A184024-1G A184024-5G
76690-658EA 8.49 USD
76690-658 76690-660
ω-Laurinlactam 99%
omega-Laurinlactam

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO
MW: 197.32 g/mol
MDL Number: MFCD00005104
CAS Number: 947-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR