Atazanavir 98%

Supplier: Ambeed
Warning

Synonyms: 1,14-Dimethyl (3S,8S,9S,12S)-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate

A182779-100MG A182779-10MG A182779-1G A182779-1MG A182779-250MG A182779-5G A182779-25G
76690-614EA 11 USD
76690-614 76690-616 76690-618 76690-620 76690-622 76690-624 77600-478
Atazanavir 98%
Atazanavir

Precaución: for Research Use Only

Formula: C₃₈H₅₂N₆O₇
MW: 704.87 g/mol
MDL Number: MFCD08435966
CAS Number: 198904-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR