Butyl-O-butyryllactate 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Butyryloxy)propionic Acid Butyl Ester, Butyl 2-(Butyryloxy)propionate, O-Butyryllactic Acid Butyl Ester, Butyl Butyryllactate

A181785-1000G A181785-100G A181785-25G A181785-500G
76690-592EA 28 USD
76690-592 76690-594 76690-596 76690-598
Butyl-O-butyryllactate 98%
Butyl-O-butyryllactate

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
MDL Number: MFCD00027213
CAS Number: 7492-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR