(±)-δ-Decalactone 97%

Supplier: Ambeed
Warning

A181647-1000G A181647-100G A181647-25G A181647-500G A181647-10G
76690-582EA 163 USD
76690-582 76690-584 76690-586 76690-588 77593-162
(±)-δ-Decalactone 97%
(±)-δ-Decalactone

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MDL Number: MFCD00006649
CAS Number: 705-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR