2-(4-(1-Oxoisoindolin-2-yl)phenyl)propanoic acid 98%

Supplier: Ambeed
Danger

Synonyms: Indoprofen

A181223-100MG A181223-10MG A181223-1MG A181223-250MG A181223-25MG A181223-50MG A181223-5G A181223-5MG
76690-560EA 40.32 USD
76690-560 76690-562 76690-564 76690-566 76690-568 76690-570 76690-572 76690-574
2-(4-(1-Oxoisoindolin-2-yl)phenyl)propanoic acid 98%
2-(4-(1-Oxoisoindolin-2-yl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₃
MW: 281.30 g/mol
MDL Number: MFCD00057144
CAS Number: 31842-01-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR