4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(5-(p-Tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulphonamide, Celecoxib
A180379-100G
A180379-10G
A180379-1G
A180379-250MG
A180379-25G
A180379-500G
A180379-5G
76690-528EA
108.22
USD
76690-528
76690-530
76690-532
76690-534
76690-536
76690-538
76690-540
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide 98%
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
Precaución: for Research Use Only
Formula:
C₁₇H₁₄F₃N₃O₂S MW: 381.38 g/mol |
MDL Number:
MFCD00941298 CAS Number: 169590-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...