7-Methoxy-3,4-dihydro-2H-benzo[b][1,4]oxazine 97%

Supplier: Ambeed
Warning

A180235-250MG A180235-5G
76690-524EA 207.94 USD
76690-524 76690-526
7-Methoxy-3,4-dihydro-2H-benzo[b][1,4]oxazine 97%
7-Methoxy-3,4-dihydro-2H-benzo[b][1,4]oxazine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD11101009
CAS Number: 93735-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR