Trifluorothymidine 98%

Supplier: Ambeed
Warning

A179915-100MG A179915-1G A179915-1MG A179915-250MG A179915-50MG A179915-5G
76690-514EA 40 USD
76690-514 76690-516 76690-518 76690-520 76690-522 77877-271
Trifluorothymidine 98%
Trifluorothymidine

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃N₂O₅
MDL Number: MFCD00006534
CAS Number: 70-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR