Benzyl phenyl sulphide 98%

Supplier: Ambeed
Danger

Synonyms: Benzyl(phenyl)sulphane

A179804-100G A179804-5G
76690-504EA 5.31 USD
76690-504 77877-268 77877-269
Benzyl phenyl sulphide 98%
Benzyl phenyl sulphide

Precaución: for Research Use Only

Formula: C₁₃H₁₂S
MDL Number: MFCD00003066
CAS Number: 831-91-4
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR