2-Methoxybenzeneboronic acid, propanediol cyclic ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Methoxyphenyl)-1,3,2-dioxaborinane

A179356-1G A179356-250MG A179356-5G
76690-498EA 10 USD
76690-498 76690-500 76690-502
2-Methoxybenzeneboronic acid, propanediol cyclic ester 98%
2-Methoxybenzeneboronic acid, propanediol cyclic ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
MDL Number: MFCD06659934
CAS Number: 141522-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR