Boc-D-Ala-OSu 97%

Supplier: Ambeed
Warning

A179113-1G A179113-250MG A179113-25G A179113-5G A179113-100G
76690-490EA 11 USD
76690-490 76690-492 76690-494 76690-496 77600-452
Boc-D-Ala-OSu 97%
Boc-D-Ala-OSu

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₆
MDL Number: MFCD00037907
CAS Number: 34404-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR