Chlorhexidine diacetate 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] diacetate, 1,1'-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Diacetate

A178012-100G A178012-10G A178012-25G A178012-500G A178012-5G
76690-440EA 28.65 USD
76690-440 76690-442 76690-444 76690-446 76690-448
Chlorhexidine diacetate 98%
Chlorhexidine diacetate

Precaución: for Research Use Only

Formula: C₂₂H₃₀Cl₂N₁₀
MW: 625.56 g/mol
MDL Number: MFCD00012532
CAS Number: 56-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR