1-(3-Bromo-4-methoxyphenyl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 3'-Bromo-4'-methoxyacetophenone, 3-Bromo-4-methoxyacetophenone

A177731-1G A177731-10G A177731-25G A177731-5G
76690-430EA 12 USD
76690-430 76690-432 77877-263 77877-264 77877-265
1-(3-Bromo-4-methoxyphenyl)ethanone 97%
1-(3-Bromo-4-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD01812494
CAS Number: 35310-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR