7-Methoxynaphthalen-1-amine 95%

Supplier: Ambeed
Warning

A177721-100MG A177721-250MG
76690-426EA 44.56 USD
76690-426 76690-428
7-Methoxynaphthalen-1-amine 95%
7-Methoxynaphthalen-1-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MW: 173.21 g/mol
MDL Number: MFCD08272073
CAS Number: 5302-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR