Cyclopentadienyltitanium trichloride 98%
Supplier: Ambeed
|
Danger
|
A177449-1G
A177449-250MG
A177449-5G
A177449-25G
76690-410EA
35
USD
76690-410
76690-412
76690-414
77625-830
Cyclopentadienyltitanium trichloride 98%
Cyclopentadienyltitanium trichloride
Precaución: for Research Use Only
Formula:
C₅Cl₃Ti |
MDL Number:
MFCD00003724 CAS Number: 1270-98-0 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...