3'-Fluoro-4'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone

A177367-1G A177367-250MG A177367-5G
76690-404EA 9.55 USD
76690-404 76690-406 76690-408
3'-Fluoro-4'-hydroxyacetophenone 98%
3'-Fluoro-4'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00215840
CAS Number: 403-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR