9,9-Bis(4-amino-3-fluorophenyl)fluorene 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(9H-Fluorene-9,9-diyl)bis(2-fluoroaniline), 2,2'-Difluoro-4,4'-(9-fluorenylidene)dianiline

A177038-10G A177038-1G A177038-25G
76690-388EA 49.87 USD
76690-388 76690-390 76690-392
9,9-Bis(4-amino-3-fluorophenyl)fluorene 97%
9,9-Bis(4-amino-3-fluorophenyl)fluorene

Precaución: for Research Use Only

Formula: C₂₅H₁₈F₂N₂
MW: 384.43 g/mol
MDL Number: MFCD08276304
CAS Number: 127926-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR