(Benzyl)triphenylphosphonium bromide 95%

Supplier: Ambeed
Warning

Synonyms: Benzyltriphenylphosphonium bromide

A176653-1000G A176653-100G A176653-10G A176653-1G A176653-25G A176653-500G
76690-376EA 77 USD
76690-376 76690-378 76690-380 76690-382 76690-384 76690-386
(Benzyl)triphenylphosphonium bromide 95%
(Benzyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₅H₂₂BrP
MW: 433.33 g/mol
MDL Number: MFCD00031707
CAS Number: 1449-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR