6-Nitro-1H-indole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A176281-100MG A176281-1G A176281-250MG A176281-5G
76690-360EA 30.77 USD
76690-360 76690-362 76690-364 76690-366
6-Nitro-1H-indole-3-carboxylic acid 95%
6-Nitro-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
MDL Number: MFCD07779489
CAS Number: 10242-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR