Arabinofuranosyl-2-fluoroadenine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoroadenine Arabinosid, (2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, Fludarabine

A175546-10MG A175546-25G A175546-50MG A175546-5G A175546-1G
76690-336EA 5.31 USD
76690-336 76690-338 76690-340 76690-342 77567-596
Arabinofuranosyl-2-fluoroadenine 98%
Arabinofuranosyl-2-fluoroadenine

Precaución: For research use only.

Formula: C₁₀H₁₂FN₅O₄
MW: 285.24 g/mol
MDL Number: MFCD00132942
CAS Number: 21679-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR