7-Methoxy-4-methylcoumarin 98%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-4-methyl-2-benzopyrone, 4-Methylherniarin

A175283-100G A175283-100MG A175283-1G A175283-250MG A175283-25G A175283-5G
76690-322EA 163.38 USD
76690-322 76690-324 76690-326 76690-328 76690-330 76690-332
7-Methoxy-4-methylcoumarin 98%
7-Methoxy-4-methylcoumarin

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
MDL Number: MFCD00009773
CAS Number: 2555-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR