5'-Bromo-2'-methoxyacetophenone 98%

Supplier: Ambeed
Warning

A175183-100G A175183-1G A175183-25G A175183-5G
76690-302EA 215 USD
76690-302 76690-304 76690-306 76690-308
5'-Bromo-2'-methoxyacetophenone 98%
5'-Bromo-2'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD03789141
CAS Number: 16740-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR