4-Bromo-1-indanone 98%
Supplier: Ambeed
|
Warning
|
A174965-100G
A174965-10G
A174965-1G
A174965-25G
A174965-500G
A174965-5G
76690-288EA
67
USD
76690-288
76690-290
76690-292
76690-294
76690-296
76690-298
4-Bromo-1-indanone 98%
4-Bromo-1-indanone
Precaución: for Research Use Only
Formula:
C₉H₇BrO |
MDL Number:
MFCD01719772 CAS Number: 15115-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...