Methyl 3-hydroxy-2-phenylpropanoate 97%

Supplier: Ambeed
Warning

A174879-10G A174879-1G A174879-250MG A174879-5G
76690-280EA 97 USD
76690-280 76690-282 76690-284 76690-286
Methyl 3-hydroxy-2-phenylpropanoate 97%
Methyl 3-hydroxy-2-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD09753766
CAS Number: 3967-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR