Crystal violet lactone 96%

Supplier: Ambeed
Warning

A173778-10G A173778-25G A173778-500G A173778-5G A173778-100G
76690-254EA 20 USD
76690-254 76690-256 76690-258 76690-260 77879-147
Crystal violet lactone 96%
Crystal violet lactone

Precaución: for Research Use Only

Formula: C₂₆H₂₉N₃O₂
MDL Number: MFCD00070611
CAS Number: 1552-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR