(±)-2-Chloromandelic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloromandelic acid, (±)-2-Chlorophenylglycolic acid

A173070-25G A173070-5G
76690-230EA 8 USD
76690-230 76690-232
(±)-2-Chloromandelic acid 97%
(±)-2-Chloromandelic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MDL Number: MFCD00084962
CAS Number: 10421-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR