9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95%

Supplier: Ambeed
Warning

A172948-1G A172948-250MG A172948-25G A172948-5G
76690-228EA 42 USD
76690-228 77877-219 77877-220 77877-221
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95%
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₃₄H₂₃BO₂
MDL Number: MFCD11045063
CAS Number: 867044-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR