7-Deaza-2'-deoxyadenosine 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3S,5R)-5-(4-Amino-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

A172863-100MG A172863-1G A172863-250MG
76690-222EA 170.81 USD
76690-222 76690-224 76690-226
7-Deaza-2'-deoxyadenosine 98%
7-Deaza-2'-deoxyadenosine

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₄O₃
MW: 250.2539 g/mol
MDL Number: MFCD00672266
CAS Number: 60129-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR