4-Chloro-2-fluorocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Chloro-2-fluorophenyl)acrylic acid

A171913-1G A171913-25G A171913-5G
76690-206EA 11.67 USD
76690-206 76690-208 76690-210
4-Chloro-2-fluorocinnamic acid 98%
4-Chloro-2-fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
MDL Number: MFCD00143287
CAS Number: 202982-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR