Diethyl-D-(+)-malate 97%

Supplier: Ambeed
Warning

A171617-1G A171617-5G A171617-100G A171617-10G
76690-182EA 5 USD
76690-182 76690-184 77625-760 77879-143
Diethyl-D-(+)-malate 97%
Diethyl-D-(+)-malate

Precaución: for Research Use Only

Formula: C₈H₁₄O₅
MDL Number: MFCD02093483
CAS Number: 7554-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR