CGS 21680 98%

Supplier: Ambeed
Warning

A170829-10MG A170829-1MG A170829-5MG A170829-50MG
76690-170EA 128 USD
76690-170 76690-172 76690-174 77625-748
CGS 21680 98%
CGS 21680

Precaución: for Research Use Only

Formula: C₂₃H₂₉N₇O₆
MDL Number: MFCD00078573
CAS Number: 120225-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR