N-(4-Aminobenzoyl)-β-alanine 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Aminobenzamido)propionic Acid, N-(4-Aminobenzoyl)-beta-alanine

A170751-100G A170751-1G A170751-5G A170751-25G
76690-164EA 133.68 USD
76690-164 76690-166 76690-168 77600-378
N-(4-Aminobenzoyl)-β-alanine 98%
N-(4-Aminobenzoyl)-beta-alanine

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
MDL Number: MFCD00009805
CAS Number: 7377-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR