3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one 95%

Supplier: Ambeed
Warning

A170265-100G A170265-100MG A170265-25G A170265-5G
76690-146EA 117 USD
76690-146 76690-148 76690-150 76690-152
3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one 95%
3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MDL Number: MFCD22123390
CAS Number: 2971-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR