2-(2-Chlorophenyl)glycine 99+%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phg(2-Cl)-OH, 2-Amino-2-(2-chlorophenyl)acetic Acid, (±)-2-Chlorophenylglycine

A170015-1000G A170015-100G A170015-10G A170015-25G A170015-500G A170015-5G
76690-134EA 87 USD
76690-134 76690-136 76690-138 76690-140 76690-142 76690-144
2-(2-Chlorophenyl)glycine 99+%
2-(2-Chlorophenyl)glycine

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD00049324
CAS Number: 88744-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR