Terephthalbis(4-fluoroaniline) 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-(1,4-Phenylene)bis(N-(4-fluorophenyl)methanimine), Bis(4-fluoroanilino)terephthalaldehyde

A168860-1G A168860-250MG
76690-122EA 48 USD
76690-122 76690-124
Terephthalbis(4-fluoroaniline) 98%
Terephthalbis(4-fluoroaniline)

Precaución: for Research Use Only

Formula: C₂₀H₁₄F₂N₂
MW: 320.34 g/mol
MDL Number: MFCD00017952
CAS Number: 17866-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR