Bendamustine hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: Bendamustine HCl, 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochloride, SDX-15 HCl, Cytostasane HCl, EP 311, 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride

A168000-100MG A168000-10MG A168000-1MG A168000-250MG A168000-50MG A168000-5MG A168000-1G
76690-104EA 25 USD
76690-104 76690-106 76690-108 76690-110 76690-112 76690-114 77877-097
Bendamustine hydrochloride 99%
Bendamustine hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₁Cl₂N₃O₂·HCl
MW: 394.72 g/mol
MDL Number: MFCD01658758
CAS Number: 3543-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR